missing translation for 'onlineSavingsMsg'
Learn More

Macherey-Nagel™ N20 PE Closures

Product Code. p-8012175 Shop All Macherey-Nagel Products
Click to view available options
Type:
Closed Top Cap
Crimp Cap
Unit Size:
Pack of 100
This item is not returnable. View return policy

Product Code. 10331102

Brand: Macherey-Nagel™ 70242

Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

Specifications

Color Transparent
Material Butyl Rubber, Polyethylene, PTFE
Septum Butyl beige / PTFE gray unassembled
Height (Metric) 9.1 mm
Quantity 100 Pk.
Thickness (Metric) 1.3 mm
Type Crimp Cap
For Use With (Equipment) Beveled HS Crimp Neck
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.