Learn More
Macherey-Nagel™ N20 Aluminum Closures
Product Code. 12480360 Shop All Macherey-Nagel Products
Click to view available options
Color:
Gold
Silver
Septum:
Butyl
Butyl/PTFE
Silicone/PTFE
Color (Seal):
Dark Gray
Dark Gray/Gray
Gray
White/Beige
Type:
Center Tear-off Crimp Cap
Complete Tear-off Crimp Cap
Crimp Cap
Pressure Release Crimp Cap
Unit Size:
Each
Pack of 100
Product Code. 12480360
Brand: Macherey-Nagel™ 70238
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Seal vials with securely with a variety of crimp closures. Machenery-Nagel™ N20 aluminum closures come with a broad variety of cap types and septum materials.
  • Center tear off caps and complete tear off caps N 20
  • Bi-metal crimp caps and magnetic crimp caps N 20
  • for use on autosamplers with magnetic transportation system –
  • Crimping of the bi-metal crimp caps is more convenient.
  • All closures mentioned above are available with silicone / PTFE septa as well as with butyl / PTFE septa in different versions.
  • Applications above 120 °C require silicone / PTFE septa.
  • Individual septa, stoppers as well as PE caps N 20 are also available.
  • Manual and electronic crimping tools N 20 complete this product range
Specifications
Color (Seal) Gray
Color Silver
Material Aluminum, Butyl Rubber
Septum Butyl
Quantity 100 Pk.
Type Complete Tear-off Crimp Cap