Learn More
Macherey-Nagel™ N20 Aluminum Closures
Product Code. 10423001 Shop All Macherey-Nagel Products
Change view
Click to view available options
Color:
Gold
Silver
Septum:
Butyl
Butyl/PTFE
Silicone/PTFE
Color (Seal):
Dark Gray
Dark Gray/Gray
Gray
White/Beige
Product Type:
Center Tear-off Crimp Cap
Complete Tear-off Crimp Cap
Crimp Cap
Pressure Release Crimp Cap
Unit Size:
Each
Pack of 100
Product Code. Color Septum Color (Seal) Product Type unitSize
10423001 Silver Butyl/PTFE Dark Gray Complete Tear-off Crimp Cap Pack of 100
10528271 Gold Butyl/PTFE Dark Gray/Gray Center Tear-off Crimp Cap Pack of 100
10072771 Silver Butyl Gray Center Tear-off Crimp Cap Pack of 100
12480360 Silver Butyl Gray Complete Tear-off Crimp Cap Pack of 100
12470360 Silver Butyl Gray Crimp Cap Pack of 100
10356651 Silver Butyl/PTFE Gray Crimp Cap Pack of 100
12715242 Silver Silicone/PTFE White/Beige Crimp Cap Each
10246283 Silver Butyl/PTFE Gray Pressure Release Crimp Cap Pack of 100
8 options
Product Code. 10423001
Brand: Macherey-Nagel™ 70235
Only null left
Add to basket
Add to basket
Seal vials with securely with a variety of crimp closures. Machenery-Nagel™ N20 aluminum closures come with a broad variety of cap types and septum materials.
  • Center tear off caps and complete tear off caps N 20
  • Bi-metal crimp caps and magnetic crimp caps N 20
  • for use on autosamplers with magnetic transportation system –
  • Crimping of the bi-metal crimp caps is more convenient.
  • All closures mentioned above are available with silicone / PTFE septa as well as with butyl / PTFE septa in different versions.
  • Applications above 120 °C require silicone / PTFE septa.
  • Individual septa, stoppers as well as PE caps N 20 are also available.
  • Manual and electronic crimping tools N 20 complete this product range
Specifications
Color (Seal) Dark Gray
Color Silver
Material Aluminum, Butyl Rubber, PTFE
Septum Butyl/PTFE
Quantity 100 Pk.
Thickness (Metric) 3 mm
Product Type Complete Tear-off Crimp Cap