missing translation for 'onlineSavingsMsg'
Learn More
Learn More
STEMCELL Technologies N2 Supplement-B
Description
- N2 Supplement-B, containing iron-poor human apo-transferrin, was developed for the in vitro differentiation of mouse or human embryonic stem (ES) cells and induced pluripotent stem (iPS) cells
- N2 Supplement-B can be used in a wide range of protocols to achieve neural induction of human ES/iPS cells and/or downstream differentiation of neural progenitor cells to different neuronal or glial subtypes
- N2 Supplement-B is provided as a 100X stock solution
Specifications
Specifications
| Cell Type | Endoderm, PSC-Derived, Neural Cells, PSC-Derived, Neural Stem and Progenitor Cells, Pancreatic Cells, Pluripotent Stem Cells |
| For Use With (Application) | Cell Culture, Differentiation |
| Quantity | 5 mL |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?