missing translation for 'onlineSavingsMsg'
Learn More

N2-Methyl-L-arginine, Tocris Bioscience™

Product Code. 18094852
Change view
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18094852 50 mg 50mg
18034212 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18094852 Supplier Tocris Bioscience Supplier No. 3513/50

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Lysosomal system c L-arginine uptake inhibitor

Selective L-arginine uptake inhibitor (Ki = 500 μM). Inhibits uptake of arginine by the lysosomal system c in human fibroblast in vitro.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2480-28-6
Molecular Formula C7H16N4O2
Molecular Weight (g/mol) 188.231
InChI Key NTWVQPHTOUKMDI-YFKPBYRVSA-N
Synonym n-methyl-l-arginine, n2-methyl-l-arginine, arginine, n2-methyl-, l, unii-88rbj4ze9r, 88rbj4ze9r, l-monomethyl-l-arginine, s-5-guanidino-2-methylamino pentanoic acid, n 2-methyl-l-arginine, n∼5∼-diaminomethylene-n∼2∼-methylornithine, n-mearg
PubChem CID 73046
IUPAC Name (2S)-5-(diaminomethylideneamino)-2-(methylamino)pentanoic acid
SMILES CNC(CCCN=C(N)N)C(=O)O

Specifications

Quantity 50 mg
Formula Weight 188.23
Percent Purity >98%
Chemical Name or Material N2-Methyl-L-arginine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.