Learn More
Macherey-Nagel™ N13 Polypropylene Black 1.3 Thickness Screw Caps
Product Code. 10187211 Shop All Macherey-Nagel Products
Change view
Click to view available options
Color:
Black
Unit Size:
Pack of 100
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Color unitSize
10187211 Black Pack of 100
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 10187211 Supplier Macherey-Nagel™ Supplier No. 702926
Only null left
Add to basket
Add to basket
Specifications
Color Black
Material Polypropylene, PTFE, Silicone
Septum Silicone White / PTFE Red
Thickness (Metric) 1.3 mm
Closure Size N13
Product Type Center