Learn More
N1-2-Thiazolylsulfanilamid, Sigma-Aldrich™
Product Code. 15804601
Change view
Click to view available options
Quantity:
250 g
Unit Size:
250g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15804601 250 g 250g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 15804601 Supplier Sigma Aldrich Supplier No. 8211120250
 Restricted Item
Estimated Shipment: 20-11-2026
For synthesis
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 72-14-0
Specifications
Melting Point 199°C to 202°C
MDL Number MFCD00005319
Quantity 250 g
Synonym N1-2-Thiazolylsulfanilamid, Sulfathiazol, N1-2-Thiazolylsulfanilamid
Molecular Weight (g/mol) 255.32
Grade Research