Learn More
N-(tert-Butyloxy)carbonyl-L-thyroxine, TRC
Specifications
Specifications
| Chemical Name or Material | N-(tert-Butyloxy)carbonyl-L-thyroxine |
| CAS | 88404-22-2 |
| Molecular Formula | C20H19I4NO6 |
| Quantity | 250 mg |
| Synonym | N-[(1,1-Dimethylethoxy)carbonyl]-O-(4-hydroxy-3,5-diiodophenyl)-3,5-diiodo-L-tyrosine |
| InChI Formula | InChI=1S/C20H19I4NO6/c1-20(2,3)31-19(29)25-15(18(27)28)6-9-4-13(23)17(14(24)5-9)30-10-7-11(21)16(26)12(22)8-10/h4-5,7-8,15,26H,6H2,1-3H3,(H,25,29)(H,27,28)/t15-/m0/s1 |
| SMILES | IC(C=C(C[C@@H](C(O)=O)NC(OC(C)(C)C)=O)C=C1I)=C1OC2=CC(I)=C(O)C(I)=C2 |
| IUPAC Name | (S)-2-((tert-butoxycarbonyl)amino)-3-(4-(4-hydroxy-3,5-diiodophenoxy)-3,5-diiodophenyl)propanoic acid |
| Molecular Weight (g/mol) | 876.99 |
| Formula Weight | 876.74 |