Learn More
n-Propyl Alcohol-d8, CDN
Specifications
Specifications
| Chemical Name or Material | 1-Propanol-D8 |
| CAS | 61393-63-3 |
| Molecular Formula | C3 D8 O |
| Quantity | 1 g |
| Synonym | 1-Propan-1,1,2,2,3,3,3-d7-ol-d (9 CI), 1,1,1,2,2,3,3-Heptadeuterio-3-deuteriooxypropane, 1-Propanol-d8, n-Propyl alcohol-d8, Propanol-d8 |
| InChI Formula | InChI=1 S/C3H8O/c1-2-3-4/h4H,2-3H2,1H3/i1D3,2D2,3D2,4 D |
| SMILES | [2 H]OC([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])[2 H] |
| IUPAC Name | 1,1,1,2,2,3,3-heptadeuterio-3-deuteriooxypropane |
| Molecular Weight (g/mol) | 68.1443 |
| Formula Weight | 68.1077 g/mol |
| Show More |