Learn More
N-Phenylpropanamide, TRC
Product Code. 30408850
Change view
Click to view available options
Quantity:
50 mg
1 g
2.5 g
Unit Size:
1g
2.5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30408850 2.5 g 2.5g
30417455 1 g 1g
30418074 50 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 30408850 Supplier Toronto Research Chemicals Supplier No. TRCP3360602.5G
 Restricted Item
Estimated Shipment: 05-08-2026
Add to basket
Add to basket
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 620-71-3
Molecular Formula C9 H11 N O
Molecular Weight (g/mol) 149.19
Synonym N-Phenylpropanamide, Fentanyl Imp. G (EP), Propanamide, N-phenyl-, Propionanilide (6CI,7CI,8CI), N-Phenylpropionamide, NSC 58952, R 50977
IUPAC Name N-phenylpropanamide
SMILES CCC(=O)Nc1ccccc1
Specifications
Quantity 2.5 g
InChI Formula InChI=1S/C9H11NO/c1-2-9(11)10-8-6-4-3-5-7-8/h3-7H,2H2,1H3,(H,10,11)
Formula Weight 149.0841
Percent Purity >95%
Chemical Name or Material N-Phenylpropanamide
Grade HPLC