Learn More
n-Pentyl-5,5,5-d3 Alcohol, CDN
Product Code. 30323628
Click to view available options
Quantity:
0.1 g
0.25 g
Unit Size:
0.1g
0.25g
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 30323628
Brand: CDN CDND52590.1G
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material 1-Pentanol-5,5,5-D3
CAS 64118-19-0
Molecular Formula C5 D3 H9 O
Quantity 0.1 g
Synonym 1-Pentan-5,5,5-d3-ol (9 CI), 1-Pentanol-5,5,5-D3, 1-Pentanol-d3, n-Pentyl-5,5,5-d3 alcohol
InChI Formula InChI=1 S/C5H12O/c1-2-3-4-5-6/h6H,2-5H2,1H3/i1D3
SMILES [2 H]C([2 H])([2 H])CCCCO
IUPAC Name 5,5,5-trideuteriopentan-1-ol
Molecular Weight (g/mol) 91.167
Formula Weight 91.108 g/mol
Percent Purity 99 atom % D, min 98% Chemical Purity