missing translation for 'onlineSavingsMsg'
Learn More

N,Nft.-Octamethylenebis(2,2-dichloroacetamide), 98.2%, MP Biomedicals™

Product Code. 11445481 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
250 mg
Unit Size:
250mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11445481 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11445481 Supplier MP Biomedicals Supplier No. 0215805083
  • ONLINE EXCLUSIVE PRICE
    1550.00 NOK
    1473.83 NOK / 250mg
    Save 76.17 NOK
    4% Off

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

A white to off-white powder used as a chemical intermediate.

Specifications

CAS 1477-57-2
Color White
Assay Percent Range 98.2%
Molecular Formula C12H20Cl4N2O2
MDL Number MFCD00000841
Quantity 250 mg
Synonym fertilysin, bis-diamine, n,n'-octane-1,8-diyl bis 2,2-dichloroacetamide, fertilysine, n,n'-octamethylenebis dichloroacetamide, bisdiamine, fertilysine n, n,n'-bis dichloroacetyl-1,8-octamethylenediamine, n,n'-octamethylenebis 2,2-dichloroacetamide, n,n'-bis dichloracetyl-1,8-octanediamine
InChI Key FAOMZVDZARKPFJ-UHFFFAOYSA-N
SMILES C(CCCCNC(=O)C(Cl)Cl)CCCNC(=O)C(Cl)Cl
IUPAC Name 2,2-dichloro-N-[8-[(2,2-dichloroacetyl)amino]octyl]acetamide
Molecular Weight (g/mol) 366.104
PubChem CID 15134
Formula Weight 366.11
Percent Purity 95%
Physical Form Powder
Chemical Name or Material N, N'-Octamethylenebis(2, 2-dichloroacetamide)
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.