Learn More
N,N,N-Trimethylethanaminium Iodide, TRC
Specifications
Specifications
| Chemical Name or Material | N,N,N-Trimethylethanaminium Iodide |
| CAS | 51-93-4 |
| Molecular Formula | C5 H14 N . I |
| Quantity | 250 mg |
| InChI Formula | InChI=1S/C5H14N.HI/c1-5-6(2,3)4;/h5H2,1-4H3;1H/q+1;/p-1 |
| SMILES | [I-].CC[N+](C)(C)C |
| IUPAC Name | ethyl(trimethyl)azanium;iodide |
| Molecular Weight (g/mol) | 215.076 |
| Formula Weight | 215.017 |