Learn More
N,N,N′,N′-Tetramethyl-1,4-phenylenediammonium Dichloride, Sigma-Aldrich™
Specifications
Specifications
| Melting Point | 227°C to 230°C (decomposition) |
| Molecular Formula | C6H4-1,4-[N(ch3)2]2 · 2 Hcl |
| MDL Number | MFCD00012482 |
| Quantity | 25 g |
| Synonym | N,N,N′,N′-Tetramethyl-1,4-phenylenediammonium dichloride, Wurster-reagent, N,N,N′,N′-Tetramethyl-1,4-phenylene-diamine dihydrochloride, N,N,N′,N′-Tetramethyl-1,4-phenylenediammonium dichloride |
| Molecular Weight (g/mol) | 237.17 |
| Grade | Research |