Learn More
N,N,N,N-Tetramethyl-1,4-phenylenediamine, 95%, BLD Pharm™
Specifications
Specifications
| CAS | 100-22-1 |
| Molecular Formula | C10H16N2 |
| MDL Number | MFCD00008309 |
| Quantity | 10 g |
| SMILES | CN(C)C1=CC=C(N(C)C)C=C1 |
| Molecular Weight (g/mol) | 164.25 |
| Percent Purity | 95% |
| Chemical Name or Material | N,N,N,N-Tetramethyl-1,4-phenylenediamine |