Learn More
N,N-Dimethylaniline-d11, TRC
Product Code. 30473871
Change view
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30473871 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 30473871 Supplier Toronto Research Chemicals Supplier No. TRCN46410210MG
 Restricted Item
Estimated Shipment: 31-08-2026
Add to basket
Add to basket
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material N,N-Dimethylaniline-d11
CAS 85785-00-8
Molecular Formula C8 D11 N
Quantity 10 mg
Synonym Benzen-d5-amine','N,N-di(methyl-d3)- (9CI), Aniline-d5','N,N-di(methyl-d3)- (6CI), N,N-Dimethylaniline-d11, N,N-Dimethylaniline-D11, N,N-Di(methyl-d3)aniline-d5
InChI Formula InChI=1S/C8H11N/c1-9(2)8-6-4-3-5-7-8/h3-7H,1-2H3/i1D3,2D3,3D,4D,5D,6D,7D
SMILES [2H]c1c([2H])c([2H])c(N(C([2H])([2H])[2H])C([2H])([2H])[2H])c([2H])c1[2H]
IUPAC Name 2,3,4,5,6-pentadeuterio-N,N-bis(trideuteriomethyl)aniline
Molecular Weight (g/mol) 132.25
Formula Weight 132.1582