Learn More
N,N-Dimethyl-p-phenylenediamine oxalate, 99%
Product Code. 10381251 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Packaging:
Glass bottle
Unit Size:
100g
25g
5g
Product Code. Quantity Packaging unitSize
10381251 25 g Glass bottle 25g
10229343 5 g Glass bottle 5g
10501814 100 g Glass bottle 100g
3 options
Product Code. 10381251
Brand: Thermo Scientific Acros 408490250
Request Bulk or Custom Format
CAS: 62778-12-5 | 0·5 C2H2O4 | 362.42 g/mol
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 62778-12-5
Molecular Formula 0·5 C2H2O4
Molecular Weight (g/mol) 362.42
MDL Number MFCD00036387
Specifications
CAS Min % 98.5
CAS Max % 100.0
Melting Point 200.0°C
Color Beige to Brown or Gray
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Glass bottle
Quantity 25 g
Formula Weight 362.42
Percent Purity 99%
Physical Form Chunks and/or Powder
Chemical Name or Material N, N-Dimethyl-p-phenylenediamine oxalate, 99%
Show More Show Less
compliance-icons
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 2
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
Hazard Statement
  • H300-Fatal if swallowed.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P330-Rinse mouth.
  • P362-Take off contaminated clothing.