missing translation for 'onlineSavingsMsg'
Learn More

N,N′-Diisopropylcarbodiimide, Sigma-Aldrich™

Product Code. 15859070
Change view
Click to view available options
Quantity:
10 mL
25 mL
Unit Size:
10mL
25mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15859070 25 mL 25mL
15849070 10 mL 10mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15859070 Supplier Sigma Aldrich Supplier No. 8036490025
 Restricted Item
Estimated Shipment: 09-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

For synthesis

N,N′-Diisopropylcarbodiimide(DIC) is a liquid carbodiimide that contains the functional group RN=C=NR. It is soluble in most organic solvents. DIC is used as a coupling agent in peptide synthesis.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 693-13-0

Specifications

Density 0.81 g/cm3 at 20°C
Boiling Point 145°C to 148°C/1013 hPa
Molecular Formula (CH3)2CHN=C=NCH(CH3)2
MDL Number MFCD00065689
Quantity 25 mL
Synonym N,N′-Diisopropylcarbodiimide, N,N′-Diisopropylcarbodiimide
Molecular Weight (g/mol) 126.2
Grade Research
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.