missing translation for 'onlineSavingsMsg'
Learn More

N,N-Diethylethylenediamine, Sigma-Aldrich™

Product Code. 15856350
Change view
Click to view available options
Quantity:
5 mL
250 mL
Unit Size:
250mL
5mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15856350 5 mL 5mL
15818950 250 mL 250mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15856350 Supplier Sigma Aldrich Supplier No. 8032150005
 Restricted Item
Estimated Shipment: 14-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

For synthesis

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 100-36-7

Specifications

Melting Point -70°C
Density 0.82 g/cm3 at 20°C
Boiling Point 145°C to 148°C/1013 hPa
Molecular Formula (C2H5)2NCH2CH2NH2
MDL Number MFCD00008176
Quantity 5 mL
Synonym N,N-Diethylethylenediamine, 2-(Diethylamino)ethylamine, 1-Amino-2-(diethylamino)ethane, N,N-Diethylethylenediamine
Molecular Weight (g/mol) 116.2
Grade Research
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.