missing translation for 'onlineSavingsMsg'
Learn More

N,N-Bis-(3-aminopropyl)methylamine, MP Biomedicals

Product Code. 11482486 Shop All MP Biomedicals Products
Click to view available options
Quantity:
10 g
Unit Size:
5g
This item is not returnable. View return policy

Product Code. 11482486

Brand: MP Biomedicals 0521528205

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Specifications

CAS 105-83-9
Molecular Formula C7H19N3
MDL Number MFCD00008217
Quantity 10 g
Synonym n,n-bis 3-aminopropyl methylamine, 3,3'-diamino-n-methyldipropylamine, bis 3-aminopropyl methylamine, n-methyliminobis propylamine, methyliminobispropylamine, methylbis 3-aminopropyl amine, 5-methyldipropylenetriamine, 1,3-propanediamine, n-3-aminopropyl-n-methyl, 3,3'-methyliminobispropylamine, n1-3-aminopropyl-n1-methylpropane-1,3-diamine
InChI Key KMBPCQSCMCEPMU-UHFFFAOYSA-N
SMILES CN(CCCN)CCCN
IUPAC Name bis(3-aminopropyl)(methyl)amine
Molecular Weight (g/mol) 145.25
PubChem CID 7777
Formula Weight 145.29
Chemical Name or Material N, N-Bis-(3-aminopropyl)methylamine
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.