Learn More
N,N'-1,2-ethanediylbis-Carbamodithioic acid C,C'-dimethyl ester, TRC
Product Code. 30132411
Change view
Click to view available options
Quantity:
2.5 mg
5 mg
10 mg
Unit Size:
10mg
2.50mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30132411 10 mg 10mg
30134512 5 mg 5mg
17982095 2.5 mg 2.50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 30132411 Supplier Toronto Research Chemicals Supplier No. TRCE50132010MG
 Restricted Item
Estimated Shipment: 02-12-2026
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 20721-48-6
Molecular Formula C6 H12 N2 S4
Molecular Weight (g/mol) 240.4329
Synonym Carbamic acid, ethylenebis[dithio-, dimethyl ester (6CI,7CI,8CI), Carbamodithioic acid, 1,2-ethanediylbis-, dimethyl ester (9CI), C,C'-Dimethyl N,N'-1,2-ethanediylbis[carbamodithioate], S,S'-Dimethylxanthogenethylenebisdithiocarbamate
IUPAC Name methyl N-[2-(methylsulfanylcarbothioylamino)ethyl]carbamodithioate
SMILES CSC(=S)NCCNC(=S)SC
Specifications
Quantity 10 mg
InChI Formula InChI=1S/C6H12N2S4/c1-11-5(9)7-3-4-8-6(10)12-2/h3-4H2,1-2H3,(H,7,9)(H,8,10)
Formula Weight 239.9883
Percent Purity >95%
Chemical Name or Material N,N'-1,2-ethanediylbis-Carbamodithioic acid C,C'-dimethyl ester
Grade HPLC