missing translation for 'onlineSavingsMsg'
Learn More

N-Methylurea, TRC

Product Code. 30481128
Change view
Click to view available options
Quantity:
5 g
25 g
50 g
Unit Size:
25g
50g
5g
Product Code. Quantity unitSize
30481128 5 g 5g
30469400 50 g 50g
30468105 25 g 25g
3 options
This item is not returnable. View return policy

Product Code. 30481128

Brand: Toronto Research Chemicals TRCM3381755G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 598-50-5
Formula Weight 74.048
InChI Formula InChI=1S/C2H6N2O/c1-4-2(3)5/h1H3,(H3,3,4,5)
IUPAC Name methylurea
Molecular Formula C2 H6 N2 O
Molecular Weight (g/mol) 74.08
SMILES CNC(=O)N
Synonym Urea, N-methyl- (ACI), N-Methylurea (ACI), Urea, methyl- (6CI, 8CI, 9CI), 1-Methylurea, MeSH ID: D008782, Methylurea, Monomethylurea
Chemical Name or Material Methylurea

Specifications

Quantity 5 g
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.