Learn More
N-(Methylsulfonyl)-4-(desnitro) Nimesulide, TRC
Specifications
Specifications
| Chemical Name or Material | N,N-Bis(methylsulphonyl)-2-phenoxyaniline |
| CAS | 905858-63-1 |
| Molecular Formula | C14 H15 N O5 S2 |
| Quantity | 25 mg |
| Synonym | N,N-Bis(methylsulphonyl)-2-phenoxyaniline, Nimesulide Imp. E (EP) |
| InChI Formula | InChI=1S/C14H15NO5S2/c1-21(16,17)15(22(2,18)19)13-10-6-7-11-14(13)20-12-8-4-3-5-9-12/h3-11H,1-2H3 |
| SMILES | CS(=O)(=O)N(c1ccccc1Oc2ccccc2)S(=O)(=O)C |
| IUPAC Name | N-methylsulfonyl-N-(2-phenoxyphenyl)methanesulfonamide |
| Molecular Weight (g/mol) | 341.4 |
| Formula Weight | 341.0392 |