Learn More
N-Methylaniline-2,3,4,5,6-d5, CDN
Product Code. 30312949
Click to view available options
Quantity:
0.1 g
0.25 g
Unit Size:
0.10g
0.25g
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 30312949
Brand: CDN CDND73680.25G
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material N-Methylaniline-2,3,4,5,6-d5
CAS 619319-96-9
Molecular Formula C7 D5 H4 N
Quantity 0.25 g
Synonym Benzen-d5-amine, N-methyl- (9 CI)
InChI Formula InChI=1 S/C7H9N/c1-8-7-5-3-2-4-6-7/h2-6,8 H,1H3/i2D,3 D,4 D,5 D,6 D
SMILES [2 H]c1c([2 H])c([2 H])c(NC)c([2 H])c1[2 H]
IUPAC Name 2,3,4,5,6-pentadeuterio-N-methylaniline
Molecular Weight (g/mol) 112.1839
Formula Weight 112.1049 g/mol
Percent Purity 99 atom % D, min 98% Chemical Purity