Learn More
n-Methyl-N-trimethylsilyl-trifluoroacetamide, Sigma-Aldrich™
Specifications
Specifications
| CAS | 24589-78-4 |
| Density | 1.085 g/cm3 |
| Boiling Point | 130°C to 132°C/1013 hPa |
| MDL Number | MFCD00000411 |
| Quantity | 5 mL |
| Synonym | N-Methyl-N-trimethylsilyl-trifluoroacetamide, MSTFA, N-Methyl-N-trimethylsilyl-trifluoroacetamide |
| Molecular Weight (g/mol) | 199.25 |
| Grade | Research |