Learn More
N-Methyl-N-(3-cyanopropyl)benzylamine, TRC
Product Code. 30483114
Change view
Click to view available options
Quantity:
250 mg
Unit Size:
250mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30483114 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 30483114 Supplier Toronto Research Chemicals Supplier No. TRCM294575250MG
 Restricted Item
Estimated Shipment: 24-09-2026
Add to basket
Add to basket
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material N-Methyl-N-(3-cyanopropyl)benzylamine
CAS 89690-05-1
Molecular Formula C12 H16 N2
Quantity 250 mg
InChI Formula InChI=1S/C12H16N2/c1-14(10-6-5-9-13)11-12-7-3-2-4-8-12/h2-4,7-8H,5-6,10-11H2,1H3
SMILES CN(CCCC#N)Cc1ccccc1
IUPAC Name 4-[benzyl(methyl)amino]butanenitrile
Molecular Weight (g/mol) 188.27
Formula Weight 188.1313