missing translation for 'onlineSavingsMsg'
Learn More

N-methyl-L-glutamic acid, MP Biomedicals™

Product Code. 11424781 Shop All MP Biomedicals Products
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
This item is not returnable. View return policy

Product Code. 11424781

Brand: MP Biomedicals 0215555583

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 6753-62-4
Molecular Formula C6H12ClNO4
Molecular Weight (g/mol) 197.615
MDL Number MFCD00065920
InChI Key LAYOOVKHWQMRIQ-WCCKRBBISA-N
Synonym n-me-glu-oh, h-l-meglu-oh hcl, n-alpha-methyl-l-glutamic acid hydrochloride, n-methyl-l-glutamic acid hydrochloride, s-2-methylamino pentanedioic acid hydrochloride
PubChem CID 56777325
IUPAC Name (2S)-2-(methylamino)pentanedioic acid;hydrochloride
SMILES CNC(CCC(=O)O)C(=O)O.Cl

Specifications

Color White
Quantity 250 mg
Formula Weight 161.16
Physical Form Solid
Chemical Name or Material N-methyl-L-glutamic acid
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.