Learn More
N-Methyl-L-alanine, TRC
Specifications
Specifications
| Chemical Name or Material | N-Methyl-L-alanine |
| CAS | 3913-67-5 |
| Molecular Formula | C4H9NO2 |
| Quantity | 1 g |
| Synonym | (S)-2-(Methylamino)propanoic Acid, (S)-N-Methylalanine, 2-(Methylamino)propanoic Acid, N-Methylalanine, alpha-(Methylamino)propionic Acid |
| InChI Formula | InChI=1S/C4H9NO2/c1-3(5-2)4(6)7/h3,5H,1-2H3,(H,6,7)/t3-/m0/s1 |
| SMILES | CN[C@@H](C)C(=O)O |
| IUPAC Name | (2S)-2-(methylamino)propanoic acid |
| Molecular Weight (g/mol) | 103.12 |
| Formula Weight | 103.0633 |