Learn More
N-Methyl-4-nitroaniline, TRC
Product Code. 30474006
Change view
Click to view available options
Quantity:
25 g
Unit Size:
25g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30474006 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 30474006 Supplier Toronto Research Chemicals Supplier No. TRCM32346025G
 Restricted Item
This item requires a price update prior to being ordered.
Request a Price for this item here.
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material N-Methyl-4-nitroaniline
CAS 100-15-2
Molecular Formula C7H8N2O2
Quantity 25 g
Synonym N-Methyl-4-nitrobenzenamine, 4-(Methylamino)nitrobenzene, 4-Nitro-N-methylaniline, Methyl(4-nitrophenyl)amine, N-(4-Nitrophenyl)-N-methylamine, NSC 5390, N-Methyl-4-nitroaniline, N-Methyl-p-nitraniline, N-Methyl-p-nitroaniline, N-Monomethyl-p-nitroaniline, p-(Methylamino)nitrobenzene, p-Nitro-N-methylaniline
InChI Formula InChI=1S/C7H8N2O2/c1-8-6-2-4-7(5-3-6)9(10)11/h2-5,8H,1H3
SMILES CNc1ccc(cc1)[N+](=O)[O-]
IUPAC Name N-methyl-4-nitroaniline
Molecular Weight (g/mol) 152.15
Formula Weight 152.0586