Learn More
N-Methyl-1-phenylmethanamine, 98%, BLD Pharm™
Specifications
Specifications
| CAS | 103-67-3 |
| Molecular Formula | C8H11N |
| MDL Number | MFCD00008289 |
| Quantity | 10 g |
| SMILES | CNCC1=CC=CC=C1 |
| Molecular Weight (g/mol) | 121.18 |
| Percent Purity | 98% |
| Chemical Name or Material | N-Methyl-1-phenylmethanamine |