Learn More
n-Hexyl-5,5,6,6,6-d5 Alcohol, CDN
Click to view available options
Quantity:
0.05 g
0.1 g
Unit Size:
0.05g
0.1g
Specifications
Specifications
| Chemical Name or Material | 1-Hexanol-5,5,6,6,6-D5 |
| CAS | 64118-18-9 |
| Molecular Formula | C6H9D5O |
| Quantity | 0.05 g |
| Synonym | 1-Hexan-5,5,6,6,6-d5-ol (9 CI), Hexyl-5,5,6,6,6-d5 alcohol, 1-Hexanol-d5, 1-Hexanol-5,5,6,6,6-D5, n-Hexyl-5,5,6,6,6-d5 alcohol, 1-Hexanol-5,5,6,6,6-d5 |
| InChI Formula | InChI=1 S/C6H14O/c1-2-3-4-5-6-7/h7H,2-6H2,1H3/i1D3,2D2 |
| SMILES | [2 H]C([2 H])([2 H])C([2 H])([2 H])CCCCO |
| IUPAC Name | hexan-5,5,6,6,6-d5-1-ol |
| Molecular Weight (g/mol) | 107.21 |
| Formula Weight | 107.14 g/mol |
| Show More |