missing translation for 'onlineSavingsMsg'
Learn More

n-Hexane, TRC

Product Code. 30136493
Change view
Click to view available options
Quantity:
100 mL
250 mL
500 mL
Unit Size:
100mL
250mL
500mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30136493 250 mL 250mL
30003585 500 mL 500mL
17962325 100 mL 100mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Regulatory: This product is under excise duties on energy product according to EU directive 2003/96/EEC.
Product Code. 30136493 Supplier Toronto Research Chemicals Supplier No. TRCH294760250ML

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: This product is under excise duties on energy product according to EU directive 2003/96/EEC.

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 110-54-3
Molecular Formula C6 H14
Molecular Weight (g/mol) 86.18
Synonym Hexane, NSC 68472, Skellysolve B, n-Hexane
IUPAC Name hexane
SMILES CCCCCC

Specifications

Quantity 250 mL
InChI Formula InChI=1S/C6H14/c1-3-5-6-4-2/h3-6H2,1-2H3
Formula Weight 86.1096
Chemical Name or Material n-Hexane
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.