Learn More
N-hexadecylphosphonicacid, 98%, BLD Pharm™
Specifications
Specifications
| CAS | 4721-17-9 |
| Molecular Formula | C16H35O3P |
| MDL Number | MFCD00015836 |
| Quantity | 100 g |
| SMILES | CCCCCCCCCCCCCCCCP(O)(O)=O |
| Molecular Weight (g/mol) | 306.42 |
| Percent Purity | 98% |
| Chemical Name or Material | N-hexadecylphosphonicacid |