missing translation for 'onlineSavingsMsg'
Learn More

n-Hexadecane, Sigma-Aldrich™

Product Code. 15876650
Change view
Click to view available options
Quantity:
5 mL
250 mL
1 L
Unit Size:
1 L
250mL
5mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15876650 1 L 1 L
15861081 5 mL 5mL
15866650 250 mL 250mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Regulatory: This product is subject to excise duty on mineral oils in accordance with European Directive 2003/96/EC.

We regret to inform you that this product is not available for the Canary Islands, Ceuta and Melilla. We apologise for the inconvenience.
Product Code. 15876650 Supplier Sigma-Aldrich Supplier No. 8206331000
 Restricted Item
Estimated Shipment: 12-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy
Regulatory: This product is subject to excise duty on mineral oils in accordance with European Directive 2003/96/EC.

We regret to inform you that this product is not available for the Canary Islands, Ceuta and Melilla. We apologise for the inconvenience.

For synthesis

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 544-76-3

Specifications

Melting Point 18°C
Density 0.77 g/cm3 at 20°C
Molecular Formula CH3(CH2)14CH3
MDL Number MFCD00008998
Quantity 1 L
Synonym n-Hexadecane, Cetane, n-Hexadecane
Molecular Weight (g/mol) 226.44
Grade Research
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.