Learn More
N-Hex-5-ynyl-2-iodo-acetamide, TRC
Click to view available options
Quantity:
50 mg
Unit Size:
50mg
Specifications
Specifications
| Chemical Name or Material | N-Hex-5-ynyl-2-iodo-acetamide |
| CAS | 930800-38-7 |
| Molecular Formula | C8H12NOI |
| Quantity | 50 mg |
| Synonym | N-5-Hexyn-1-yl-2-iodoacetamide |
| InChI Formula | InChI=1S/C8H12INO/c1-2-3-4-5-6-10-8(11)7-9/h1H,3-7H2,(H,10,11) |
| SMILES | ICC(=O)NCCCCC#C |
| IUPAC Name | N-hex-5-ynyl-2-iodoacetamide |
| Molecular Weight (g/mol) | 265.09 |
| Formula Weight | 264.9964 |