Learn More
n-Heptylbenzene-d20, CDN
Click to view available options
Quantity:
0.5 g
1 g
Unit Size:
0.5g
1g
Specifications
Specifications
| Chemical Name or Material | Heptylbenzene-D20 |
| CAS | 634897-84-0 |
| Molecular Formula | C13 D20 |
| Quantity | 1 g |
| Synonym | Benzene-d5, heptyl-d15- (9 CI), n-Heptylbenzene-d20, Heptylbenzene-D20, Heptyl-d15-benzene-d5 |
| InChI Formula | InChI=1 S/C13H20/c1-2-3-4-5-7-10-13-11-8-6-9-12-13/h6,8-9,11-12 H,2-5,7,10H2,1H3/i1D3,2D2,3D2,4D2,5D2,6 D,7D2,8 D,9 D,10D2,11 D,12 D |
| SMILES | [2 H]c1c([2 H])c([2 H])c(c([2 H])c1[2 H])C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])[2 H] |
| IUPAC Name | 1,2,3,4,5-pentadeuterio-6-(1,1,2,2,3,3,4,4,5,5,6,6,7,7,7-pentadecadeuterioheptyl)benzene |
| Molecular Weight (g/mol) | 196.421 |
| Formula Weight | 196.282 g/mol |
| Show More |