Learn More
n-Heptadecane, Sigma-Aldrich™
Specifications
Specifications
| CAS | 629-78-7 |
| Melting Point | 22°C |
| Density | 0.78 g/cm3 at 20°C |
| Boiling Point | 302°C/1013 hPa |
| Molecular Formula | CH3(CH2)15CH3 |
| MDL Number | MFCD00009002 |
| Quantity | 50 mL |
| Synonym | n-Heptadecane, n-Heptadecane |
| Molecular Weight (g/mol) | 240.47 |
| Grade | Research |