missing translation for 'onlineSavingsMsg'
Learn More

N-Fmoc-L-homophenylalanine, 95%, Thermo Scientific Chemicals

Product Code. 15408447
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
This item is not returnable. View return policy

Product Code. 15408447

Brand: Thermo Scientific Alfa Aesar H51973.03

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

It is used as an active pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used as an active pharmaceutical intermediate.

Solubility
Slightly soluble in water.

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 132684-59-4
Molecular Formula C25H23NO4
Molecular Weight (g/mol) 401.46
MDL Number MFCD00077046
InChI Key CIHPCIUGLIZADU-QHCPKHFHSA-N
Synonym fmoc-l-homophenylalanine, fmoc-homophe-oh, fmoc-hophe-oh, fmoc-l-homophenyl-ala, fmoc-l-homophe, fmoc-l-hphe-oh, n-fmoc-l-homophenylalanine, s-2-9h-fluoren-9-yl methoxy carbonyl amino-4-phenylbutanoic acid, s-2-fmoc-amino-4-phenylbutyric acid, 2s-2-9h-fluoren-9-ylmethoxy carbonyl amino-4-phenylbutanoic acid
PubChem CID 2761467
IUPAC Name (2S)-2-(9H-fluoren-9-ylmethoxycarbonylamino)-4-phenylbutanoic acid
SMILES OC(=O)[C@H](CCC1=CC=CC=C1)NC(=O)OCC1C2=CC=CC=C2C2=CC=CC=C12

Specifications

Quantity 1 g
Beilstein 4847669
Solubility Information Slightly soluble in water.
Formula Weight 401.46
Percent Purity 95%
Chemical Name or Material N-Fmoc-L-homophenylalanine

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.