missing translation for 'onlineSavingsMsg'
Learn More

n-Eicosane, Sigma-Aldrich™

Product Code. 15881021
Change view
Click to view available options
Quantity:
50 g
Unit Size:
50g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15881021 50 g 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Regulatory: This product is subject to excise duty on mineral oils in accordance with European Directive 2003/96/EC.

We regret to inform you that this product is not available for the Canary Islands, Ceuta and Melilla. We apologise for the inconvenience.
Product Code. 15881021 Supplier Sigma Aldrich Supplier No. 8205470050
 Restricted Item
Estimated Shipment: 16-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy
Regulatory: This product is subject to excise duty on mineral oils in accordance with European Directive 2003/96/EC.

We regret to inform you that this product is not available for the Canary Islands, Ceuta and Melilla. We apologise for the inconvenience.

For synthesis

TRUSTED_SUSTAINABILITY

Specifications

CAS 112-95-8
Melting Point 34°C to 37°C
Density 0.79 g/cm3 at 20°C
Boiling Point 204°C to 206°C/20 hPa
Molecular Formula CH3(CH2)18CH3
MDL Number MFCD00009344
Quantity 50 g
Synonym n-Eicosane, n-Eicosane
Molecular Weight (g/mol) 282.55
Grade Research
compliance-icons
Product Identifier
  • n-Eicosane for synthesis
Signal Word
  • Danger
Hazard Category
  • Aspiration toxicity Category 1
  • Specific target organ toxicity Category 3
Hazard Statement
  • H304-May be fatal if swallowed and enters airways.
Precautionary Statement
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P331-Do NOT induce vomiting.
  • P405-Store locked up.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
  • MIXTURE LIST-Contain: Isocane
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.