Learn More
n-Dodecylpyridinium Chloride, Sigma-Aldrich™
Product Code. 15871021
Change view
Click to view available options
Quantity:
5 g
250 g
1 kg
Unit Size:
1kg
250g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15871021 1 kg 1kg
15851021 5 g 5g
15861021 250 g 250g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 15871021 Supplier Sigma Aldrich Supplier No. 8205461000
 Restricted Item
Estimated Shipment: 26-11-2026
For synthesis
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 104-74-5
Specifications
Melting Point 87°C to 89°C
Density 1.04 g/cm3 at 20°C
Molecular Formula CH3(CH2)11C5H5NCl
MDL Number MFCD00011987
Quantity 1 kg
Synonym N-Dodecylpyridinium chloride, N-Laurylpyridinium chloride, N-Dodecylpyridinium chloride
Molecular Weight (g/mol) 283.88
Grade Research