missing translation for 'onlineSavingsMsg'
Learn More

n-Dodecane, Sigma-Aldrich™

Product Code. 15886640
Change view
Click to view available options
Quantity:
100 mL
250 mL
Unit Size:
100mL
250mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15886640 250 mL 250mL
15876640 100 mL 100mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Regulatory: This product is subject to excise duty on mineral oils in accordance with European Directive 2003/96/EC.

We regret to inform you that this product is not available for the Canary Islands, Ceuta and Melilla. We apologise for the inconvenience.
Product Code. 15886640 Supplier Sigma Aldrich Supplier No. 8205430250
 Restricted Item
Estimated Shipment: 14-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy
Regulatory: This product is subject to excise duty on mineral oils in accordance with European Directive 2003/96/EC.

We regret to inform you that this product is not available for the Canary Islands, Ceuta and Melilla. We apologise for the inconvenience.

For synthesis

n-Dodecane is used as a solvent in organic chemistry. It is used as a distillation chaser and scintillator component. Currently, n-dodecane is being considered an alternative to kerosene-based fuels.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 112-40-3

Specifications

Melting Point -9.6°C
Density 0.75 g/cm3 at 20°C
Boiling Point 216.3°C/1013 hPa
Molecular Formula CH3(CH2)10CH3
MDL Number MFCD00008969
Quantity 250 mL
Synonym n-Dodecane, n-Dodecane
Molecular Weight (g/mol) 170.33
Grade Research
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.