Learn More
n-Dodecane, MP Biomedicals™
Product Code. 11488302 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
2.5 L
Unit Size:
2.5 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11488302 2.5 L 2.5 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 11488302 Supplier MP Biomedicals Supplier No. 0219515896
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Used as a solvent, distillation chaser and scintillator component. n-Dodecane, MP Biomedicals is a liquid alkane hydrocarbon that is also used as a diluent for tributyl phosphate (TBP) in plants reprocessing.
Specifications
CAS 112-40-3
Melting Point -9.6°C (literature)
Density 0.753 at 25°C (water = 1) (Lit.)
Boiling Point 215°C to 217°C (literature)
Molecular Formula C12H26
Refractive Index n20/D 1.421 (Lit.)
Quantity 2.5 L
Synonym n-dodecane, dihexyl, bihexyl, adakane 12, n-dodecan, n-dodecan german, duodecane, alkanes, c10-14, undecane, methyl, ccris 661
InChI Key SNRUBQQJIBEYMU-UHFFFAOYSA-N
SMILES CCCCCCCCCCCC
IUPAC Name dodecane
Molecular Weight (g/mol) 170.34
PubChem CID 8182
ChEBI CHEBI:28817
Formula Weight 170.34g/mol
Physical Form Liquid