Learn More
N-Desmethyllevomepromazine Hydrochloride, LoGiCal
Product Code. 30463477
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
Product Code. 30463477
Brand: LoGiCal LGCFOR0019.07
LoGiCal's certified reference materials as neats, native solutions and stable isotope labelled solutions, with expert documentation and support. From forensic to clinical labs, choose LoGiCal for confidence, compliance, and analytical excellence.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material N-Desmethyllevomepromazine Hydrochloride
CAS 61733-92-4
Analyte or Component Names N-Desmethyllevomepromazine Hydrochloride
Shipping Condition Room Temperature
Molecular Formula C18 H22 N2 O S . Cl H
Certifications/Compliance ISO 9001
Synonym N-Demethyllevomepromazine Hydrochloride
InChI Formula InChI=1S/C18H22N2OS.ClH/c1-13(11-19-2)12-20-15-6-4-5-7-17(15)22-18-9-8-14(21-3)10-16(18)20;/h4-10,13,19H,11-12H2,1-3H3;1H/t13-;/m1./s1
Solution Type Neat
SMILES Cl.CNC[C@@H](C)CN1c2ccccc2Sc3ccc(OC)cc13
IUPAC Name (2R)-3-(2-methoxyphenothiazin-10-yl)-N,2-dimethylpropan-1-amine;hydrochloride
Industry Type Clinical Testing
Molecular Weight (g/mol) 350.91
Quantity 10 mg
Formula Weight 350.122 g/mol