missing translation for 'onlineSavingsMsg'
Learn More

n-Decane, MP Biomedicals™

Product Code. 11478302 Shop All MP Biomedicals Products
Click to view available options
Quantity:
100 mL
500 mL
2.5 L
Unit Size:
100mL
2.5 L
500mL
This item is not returnable. View return policy

Product Code. 11478302

Brand: MP Biomedicals 0219512396

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

n-Decane >99%, Clear, Colorless Liquid

  • Decane has been used as a solvent in the preparation of high alkane phase emulsified suspensions (HAPES).
  • It may also be used as a solvent to suspend the CdSe/ZnS nanocrystals during absorbance measurement.
  • Internal standard in the GC analysis of oxalic, malonic, and succinic acids in biological materials.
  • Decane (n-Decane), an aliphatic hydrocarbon, is a reference fuel. Its oxidation over a range of temperature has been studied.
  • Its crystal structure has been investigated at 150K.
  • The cracking of decane can be performed using electrohydrodynamic atomization system (EHDA).
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 124-18-5
Molecular Formula C10H22
Molecular Weight (g/mol) 142.29
InChI Key DIOQZVSQGTUSAI-UHFFFAOYSA-N

Specifications

Melting Point -30°C (literature)
Density 0.734 g/cm3 at 15°C(Lit.)
Boiling Point 174°C (Literature)
Refractive Index n20/D 1.411(Lit.)
Quantity 2.5 L
UN Number UN1760
Percent Purity >99%
Physical Form Liquid
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.