Learn More
N-deacetylated BMS-202, MedChemExpress
Product Code. 16497878
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
25 mg
50 mg
100 mg
Unit Size:
100mg
10mg
25mg
50mg
5mg
Product Code. 16497878
Brand: Medchem Express HY19745A
MedChemExpress N-deacetylated BMS-202 is the deacetylated of BMS-202. BMS-202 is an inhibitor of the PD-1/PD-L1 interaction, mainly used for cancer treatment.
N-deacetylated BMS-202 is the deacetylated of BMS-202. BMS-202 is an inhibitor of the PD-1/PD-L1 interaction, mainly used for cancer treatment.
Specifications
Chemical Name or Material N-deacetylated BMS-202
Physical Form Solid
CAS 2310135-18-1
Color Yellow
Quantity 10 mg
Purity Grade Notes Research
Molecular Formula C23H27N3O2
For Use With (Application) Cancer-programmed cell death
Solubility Information DMSO : ≥ 36 mg/mL (95.37 mM)
SMILES NCCNCC1=CC=C(OCC2=C(C)C(C3=CC=CC=C3)=CC=C2)N=C1OC
Molecular Weight (g/mol) 377.48
Formula Weight 377.48
Percent Purity 98.13%
Grade Research
Show More Show Less
Research purposes only
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.