Learn More
N-Deacetyl-N-formyl Colchicine, TRC
Product Code. 30007883
Change view
Click to view available options
Quantity:
10 mg
25 mg
50 mg
Packungsgröße:
10mg
25mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Quantity unitSize
30007883 25 mg 25mg
30011435 50 mg 50mg
30003282 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
Artikelnummer. 30007883 Lieferant Toronto Research Chemicals Lieferanten-Nr. TRCD19894025MG
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemische Identifikatoren
CAS 7411-12-3
Molecular Formula C21 H23 N O6
Molecular Weight (g/mol) 385.41
Synonym N-[(7S,12aRa)-1,2,3,10-tetramethoxy-9-oxo-5,6,7,9-tetrahydrobenzo[a]heptalen-7-yl]formamide, N-[(7S)-5,6,7,9-Tetrahydro-1,2,3,10-tetramethoxy-9-oxobenzo[a]heptalen-7-yl]-formamide, Colchicine, N-deacetyl-N-formyl- (7CI,8CI), Colchicine, deacetyl-N-formyl- (6CI), Formamide, N-(5,6,7,9-tetrahydro-1,2,3,10-tetramethoxy-9-oxobenzo[a]heptalen-7-yl)-, (S)-, Benzo[a]heptalene, formamide deriv., Gloriosine, N-Deacetyl-N-formylcolchicine, N-Formyl-N-deacetylcolchicine, N-Formyl-N-desacetylcolchicine, N-Formyldeacetylcolchicine, NSC 403142, Colchicine Imp. A (EP)
IUPAC Name N-[(7S)-1,2,3,10-tetramethoxy-9-oxo-6,7-dihydro-5H-benzo[a]heptalen-7-yl]formamide
SMILES COC1=CC=C2C(=CC1=O)[C@H](CCc3cc(OC)c(OC)c(OC)c23)NC=O
Spezifikation
Quantity 25 mg
InChI Formula InChI=1S/C21H23NO6/c1-25-17-8-6-13-14(10-16(17)24)15(22-11-23)7-5-12-9-18(26-2)20(27-3)21(28-4)19(12)13/h6,8-11,15H,5,7H2,1-4H3,(H,22,23)/t15-/m0/s1
Formula Weight 385.1525
Percent Purity >95%
Chemical Name or Material N-Deacetyl-N-formyl Colchicine
Grade HPLC