Learn More
N-Carbamimidoylacetamide, 97%, BLD Pharm™
Specifications
Specifications
| CAS | 5699-40-1 |
| Molecular Formula | C3H7N3O |
| MDL Number | MFCD00075615 |
| Quantity | 5 g |
| SMILES | CC(=O)NC(N)=N |
| Molecular Weight (g/mol) | 101.11 |
| Percent Purity | 97% |
| Chemical Name or Material | N-Carbamimidoylacetamide |