missing translation for 'onlineSavingsMsg'
Learn More
Learn More
N-Cadherin Mouse anti-Human, Alexa Fluor 647, Clone: 691723, R&D Systems™

Mouse Monoclonal Antibody
Brand: R&D Systems FAB13881R-100UG
This item is not returnable.
View return policy
Description
N-Cadherin Monoclonal specifically detects N-Cadherin in Human samples. It is validated for Flow Cytometry.Specifications
| N-Cadherin | |
| Monoclonal | |
| 0.2 mg/mL | |
| Flow Cytometry 0.25-1 ug/10^6 cells | |
| P19022.4 | |
| CDH2 | |
| Mouse myeloma cell line NS0-derived recombinant mouse N-Cadherin Asp160-Ala724 Accession # P19022.4 | |
| RUO | |
| 1000 | |
| Human | |
| IgM |
| Flow Cytometry | |
| 691723 | |
| Alexa Fluor 647 | |
| Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. with 0.09% Sodium Azide | |
| ACOGS, ARVD14, cadherin 2, type 1, N-cadherin (neuronal), cadherin-2, calcium-dependent adhesion protein, neuronal, CD325, CD325 antigen, CDH2, CDHN, CDw325, NCAD, N-cadherin, NCadherin, N-cadherin 1, Neural cadherin, neural-cadherin | |
| Mouse | |
| 100 μg | |
| Primary | |
| Detects mouse N-Cadherin in direct ELISAs and Western blots. In direct ELISAs approximately 50% cross-reactivity with recombinant mouse N-Cadherin is observed, and no cross-reactivity with recombinant human (rh) E-Cadherin, rhP-Cadherin, rhVE-Cadherin, rhCadherin-4, -8, -11, -12, or -13 is observed. | |
| Store the unopened product at 2 - 8 degreesC. Do not use past expiration date. |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
For Research Use Only (RUO)
Spot an opportunity for improvement?Share a Content Correction