Learn More
n-Butylethylmagnesium, 0.9M solution in heptane, AcroSeal™
Product Code. 10778833 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
100 mL
800 mL
Packaging:
AcroSeal™ Glass Bottle
Unit Size:
100mL
800mL
Product Code. 10778833
Brand: Thermo Scientific Acros 381188000
Request Bulk or Custom Format
CAS: 62202-86-2 | C6H14Mg | 110.48 g/mol
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 62202-86-2, 142-82-5
Molecular Formula C6H14Mg
Molecular Weight (g/mol) 110.48
MDL Number MFCD00072569
InChI Key MVECFARLYQAUNR-UHFFFAOYSA-N
Synonym magnesium, butylethyl, butylethylmagnesium, n-butylethylmagnesium, butyl ethyl magnesium, n-butyl ethyl magnesium, mvecfarlyqaunr-uhfffaoysa-n
PubChem CID 112818
IUPAC Name magnesium;butane;ethane
SMILES CCCC[Mg]CC
Specifications
Color Colorless
Density 0.7300g/mL
Packaging AcroSeal™ Glass Bottle
Linear Formula CH3CH2Mg(CH2)3CH3
Quantity 800 mL
Specific Gravity 0.73
Solubility Information Solubility in water: reacts
Formula Weight 110.48
Physical Form Liquid
Chemical Name or Material n-Butylethylmagnesium, 0.9M Solution in Heptane