missing translation for 'onlineSavingsMsg'
Learn More

n-Butyl Acetate, Supelco™

Product Code. 15897050
Change view
Click to view available options
Quantity:
2.5 L
25 L
190 L
Unit Size:
190 L
2.5 L
25 L
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15897050 190 L 190 L
15844160 2.5 L 2.5 L
15854160 25 L 25 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15897050 Supplier Supelco Supplier No. 1019749190
 Restricted Item
Estimated Shipment: 16-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

EMPLURA™

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 123-86-4

Specifications

Melting Point −78°C (lit.)
Density 0.88 g/mL at 25°C
Boiling Point 124°C to 126°C
Molecular Formula CH3COO(CH2)3CH3
MDL Number MFCD00009445
Quantity 190 L
Synonym Butyl acetate, Acetic acid n-butyl ester, Butyl ethanoate, Butyl acetate
Molecular Weight (g/mol) 116.16
Grade Research
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.